Kaliumhydrogendifluorid ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Kaliumhydrogenfluorid, Kaliumbifluorid
A13031.22
A13031.0E
A13031.36
A13031.22EA
28.4
EUR
A13031.22
A13031.0E
A13031.36
Kaliumhydrogendifluorid ≥98%
Potassium hydrogen difluoride
Purity: 98%
CAS No.: 7789-29-9
Molecular Formula: KHF2
Formula Weight: 78.10
Odour: Odourless
pH: 1
Melting Point: 239° dec.
Merck Reference: 14,7610
UN No.: 1811
MDL No.: MFCD00011403
Formel:
HF₂K Molecular Weight: 78,1 g/mol Schmelzpunkt: 239 °C (Zersetzung) Dichte: 2,370 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011403 CAS-Nummer: 7789-29-9 EINECS: 232-156-2 UN: 1811 ADR: 8,II Merck Index: 14,7610 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...