Bismutnatriumtrioxid ≥80% ACS
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Natriumbismutat(V), Sodium bismuthate (V), Natriumtrioxobismutat(V)
33219.14
33219.22
33219.14EA
85.7
EUR
33219.14
33219.22
Bismutnatriumtrioxid ≥80% ACS
Bismutnatriumtrioxid
Purity: 80% min
CAS No.: 12232-99-4
Molecular Formula: NaBiO3
Formula Weight: 279.97
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: Yellow to brown
Odour: Odourless
Solubility: Insoluble in cold water. Decomposes in hot water forming Bi2O3, NaOH, and oxygen
Merck Reference: 14,8585
MDL No.: MFCD00003474
Manganese testing in iron, steel, ores
Formel:
BiNaO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003474 CAS-Nummer: 12232-99-4 EINECS: 235-455-6 Merck Index: 14,8585 |