Kaliumcarbonat Sesquihydrat 98,5-101,0% ACS
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Pottasche Sesquihydrat
10981.22
10981.36
10981.22EA
37.7
EUR
10981.22
10981.36
Kaliumcarbonat Sesquihydrat 98,5-101,0% ACS
Kaliumcarbonat Sesquihydrat
Purity: 98.5-101.0%
CAS No.: 6381-79-9
Molecular Formula: K2CO3.1.5H2O
Formula Weight: 165.23 (138.21anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
Solubility: Soluble in water. Practically insoluble in alcohol
Merck Reference: 14,7670
MDL No.: MFCD00149925
Manufacturing of soaps, pigments, printing ink, glass, pottery, other potassium salts, as an analytical lab reagent
Formel:
CK₂O₃·1,5H₂O Molecular Weight: 165,23 g/mol Schmelzpunkt: 891 °C Dichte: 2,29 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149925 CAS-Nummer: 6381-79-9 EINECS: 209-529-3 Merck Index: 14,7670 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...