Kaliumhexafluorosilicat ≥99,999% (Metall-Basis)
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Potassium fluorosilicate
44495.09
44495.18
44495.30
44495.09EA
67.4
EUR
44495.09
44495.18
44495.30
Kaliumhexafluorosilicat ≥99,999% (Metall-Basis)
Potassium hexafluorosilicate
Purity: 99.999%
CAS No.: 16871-90-2
Molecular Formula: F6K2Si
Formula Weight: 220.28
Physical Form: Crystalline
Density: 2.27
Appearance: White
pH: 3.4
Melting Point: dec.
Merck Reference: 14,7638
UN No.: 2655
MDL No.: MFCD00040666
Formel:
F₆K₂Si Molecular Weight: 220,27 g/mol Schmelzpunkt: Zersetzung Dichte: 2,27 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00040666 CAS-Nummer: 16871-90-2 EINECS: 240-896-2 UN: 2655 ADR: 6.1,III Merck Index: 14,7638 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...