Gallium(III)iodid, ultra trocken ≥99,999% (Metall-Basis)
Lieferant: Thermo Scientific
|
Gefahr
|
Unter Argon in Ampullen verpackt
35699.03
35699.06
35699.14
35699.03EA
74.3
EUR
35699.03
35699.06
35699.14
Gallium(III)iodid, ultra trocken ≥99,999% (Metall-Basis)
Gallium triiodide
Purity: 99.999%
CAS No.: 13450-91-4
Molecular Formula: GaI3
Formula Weight: 450.43
Physical Form: Powder, Ampouled under argon
Density: 4.15
Appearance: Yellow-green
Melting Point: 212°
Boiling Point: 345° subl.
UN No.: 3260
MDL No.: MFCD00016107
Formel:
GaI₃ Molecular Weight: 450,44 g/mol Siedepunkt: 345 °C (Subl.) Schmelzpunkt: 212 °C Dichte: 4,15 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016107 CAS-Nummer: 13450-91-4 EINECS: 236-611-6 UN: 3260 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...