Digalliumtrioxid ≥99,999% (Metall-Basis), Puratronic®

Lieferant: Thermo Scientific

11151.22EA 787 EUR
11151.22 11151.06 11151.36 11151.14
Digalliumtrioxid ≥99,999% (Metall-Basis), Puratronic®
Digalliumtrioxid
Formel: Ga₂O₃
Molecular Weight: 187,44 g/mol
Schmelzpunkt: 1900 °C
Dichte: 6,44
Lagertemperatur: Raumtemperatur
MDL: MFCD00011020
CAS-Nummer: 12024-21-4
EINECS: 234-691-7
Merck Index: 14,4346

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR