Indium(III)acetylacetonat ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Indium(III)-2,4-pentandionat, Tris(2,4-pentandionato-O,O')indium(III), Indium(III)-tris(2,4-pentandionato-O,O'), Tri(2,4-pentandionato-O,O')indium(III)
13794.03
13794.06
13794.14
13794.03EA
44.2
EUR
13794.03
13794.06
13794.14
Indium(III)acetylacetonat ≥98%
Indium(III)acetylacetonat
Purity: 98%
CAS No.: 14405-45-9
Molecular Formula: C15H21InO6
Formula Weight: 412.15
Physical Form: Powder
Melting Point: 186° dec.
MDL No.: MFCD00013494
Used in catalysts
Formel:
C₁₅H₂₁InO₆ Molecular Weight: 412.15 g/mol Schmelzpunkt: 186 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013494 CAS-Nummer: 14405-45-9 EINECS: 238-378-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...