Diindiumtrisulfat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Diindiumtris(sulfat), Indium(III)sulfat

57151-10G 57151-50G
SIAL57151-10GEA 189 EUR
SIAL57151-10G SIAL57151-50G
Diindiumtrisulfat, Sigma-Aldrich®
Diindiumtrisulfat
Formel: O₁₂In₂S₃
Molecular Weight: 517,83 g/mol
Schmelzpunkt: 585 °C
Dichte: 3,438 g/cm³ (20 °C)
MDL: MFCD00011059
CAS-Nummer: 13464-82-9
EINECS: 236-689-1
Merck Index: 13,04978

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR