Cyclohexyltrichlorsilan

Lieferant: CHEMPUR SILANE

Synonyme: Cyclohexylsilyltrichlorid, Trichlor(cyclohexyl)silan

SILS05225.10 SILS05225.25 SILS05225.100
SILNSILS05225.10EA 0 EUR
SILNSILS05225.10 SILNSILS05225.25 SILNSILS05225.100
Cyclohexyltrichlorsilan
Cyclohexyltrichlorsilan
Formel: C₆H₁₁Cl₃Si
Molecular Weight: 217.6 g/mol
Siedepunkt: 81
Dichte: 1.232 g/cm³ (20 °C)
MDL: MFCD00021280
CAS-Nummer: 98-12-4
EINECS: 202-639-2
UN: 1763
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR