Bismut(III)chloridoxid ≥99,999% (Metall-Basis)

Lieferant: Thermo Scientific
Achtung

Synonyme: Bismutoxychlorid

11843.09 11843.18
11843.09EA 79.9 EUR
11843.09 11843.18
Bismut(III)chloridoxid ≥99,999% (Metall-Basis)
Bismuth chloride oxide

Purity: 99.999% CAS No.: 7787-59-9 Molecular Formula: BiClO Formula Weight: 260.43 Physical Form: Powder Density: 7.72 Refractive Index: 2.15 Appearance: Very pale brown Odour: Odourless Melting Point: dec. Merck Reference: 14,1263 MDL No.: MFCD00010895

Formel: BiClO
Molecular Weight: 260,43 g/mol
Schmelzpunkt: Zersetzung
Dichte: 7,72
Lagertemperatur: Raumtemperatur
MDL: MFCD00010895
CAS-Nummer: 7787-59-9
EINECS: 232-122-7
Merck Index: 14,1263

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR