Tetradecyltrimethylammoniumchlorid ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Myristyltrimethylammoniumchlorid, Tetradoniumchlorid, TTAC

L12484.06 L12484.14
L12484.06EA 42.1 EUR
L12484.06 L12484.14
Tetradecyltrimethylammoniumchlorid ≥98%
Tetradecyltrimethylammoniumchlorid

Purity: 98% CAS No.: 4574-04-3 Molecular Formula: CH3(CH2)13N(CH3)3Cl Formula Weight: 291.95 Beilstein Reference: 3916572 MDL No.: MFCD00059968

Formel: C₁₇H₃₈ClN
Molecular Weight: 291,95 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00059968
CAS-Nummer: 4574-04-3
EINECS: 224-958-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR