Tetrabutylammoniumhexafluorophosphat

Lieferant: FLUOROCHEM

Synonyme: Tetrabutylammonium hexafluorophosphate(V), Tetra-n-butylammonium hexafluorophosphate

007314-25G 007314-100G
FLUO007314-25GEA 67.1 EUR
FLUO007314-25G FLUO007314-100G
Tetrabutylammoniumhexafluorophosphat
Tetrabutylammonium hexafluorophosphate
Formel: C₁₆H₃₆F₆NP
Molecular Weight: 387,43 g/mol
MDL: MFCD00011748
CAS-Nummer: 3109-63-5
EINECS: 221-472-6
UN: 3263
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR