Ammoniummolybdat(VI), Sigma-Aldrich®

Lieferant: Merck

Synonyme: Molybdic acid ammonium salt, Molybdic acid, ammonium salt, Ammonium molybdate, di-Ammonium molybdate

M-Clarity™ Qualitätsstufe = MQ100

277908-100G 277908-20G 277908-5G
SIAL277908-100GEA 1410 EUR
SIAL277908-100G SIAL277908-20G SIAL277908-5G
Ammoniummolybdat(VI), Sigma-Aldrich®
Ammonium molybdate(VI)
Formel: H₈MoN₂O₄
Molecular Weight: 196,01 g/mol
MDL: MFCD00011596
CAS-Nummer: 13106-76-8
EINECS: 236-031-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR