Benzylphenylsulfid
Lieferant: FLUOROCHEM
Synonyme:
S-Benzylthiophenol
003349-100G
003349-25G
FLUO003349-100GEA
210
EUR
FLUO003349-100G
FLUO003349-25G
Benzylphenylsulfid
Benzylphenylsulfid
Formel:
C₁₃H₁₂S Molecular Weight: 200,3 g/mol Siedepunkt: 197 °C (1013 hPa) Schmelzpunkt: 40…44 °C |
MDL:
MFCD00003066 CAS-Nummer: 831-91-4 EINECS: 212-612-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...