Certrichlorid
Lieferant: CHEMPUR
|
Achtung
|
Synonyme:
Cer(III)chlorid
900845.0025
900845
CHMP900845.0025EA
0
EUR
CHMP900845.0025
CHMP900845
Certrichlorid
Certrichlorid
Formel:
CeCl₃ Molecular Weight: 246,47 g/mol Schmelzpunkt: 848 °C Dichte: 3,193 g/cm³ (20 °C) |
MDL:
MFCD00010929 CAS-Nummer: 7790-86-5 EINECS: 232-227-8 Merck Index: 13,02008 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...