2-Hydroxy-1-naphthaldehyd, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Hydroxy-α-naphthaldehyde

M-Clarity™ Qualitätsstufe = MQ100

H45353-100G H45353-25G
SIALH45353-100GEA 66.5 EUR
SIALH45353-100G SIALH45353-25G
2-Hydroxy-1-naphthaldehyd, Sigma-Aldrich®
2-Hydroxy-1-naphthaldehyd
Formel: C₁₁H₈O₂
Molecular Weight: 172,18 g/mol
MDL: MFCD00004005
CAS-Nummer: 708-06-5
EINECS: 211-902-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR