N,N-Dimethyl-1-naphthylamin ≥99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Dimethyl(1-naphthyl)amin
36438.AA
36438.AC
36438.AAEA
37.1
EUR
36438.AA
36438.AC
N,N-Dimethyl-1-naphthylamin ≥99%
N,N-Dimethyl-1-naphthylamin
Purity: 99%
CAS No.: 86-56-6
Molecular Formula: C10H7N(CH3)2
Formula Weight: 171.24
Physical Form: Liquid
Density: 1.039
Refractive Index: 1.6230
Appearance: Pale yellow
Boiling Point: 145-147°/12mm
Merck Reference: 14,3252
UN No.: 3082
MDL No.: MFCD00003919
Formel:
C₁₂H₁₃N Molecular Weight: 171,24 g/mol Siedepunkt: 145…147 °C (12 mmHg) Dichte: 1,039 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003919 CAS-Nummer: 86-56-6 EINECS: 201-682-4 UN: 3082 ADR: 9,III Merck Index: 14,3252 |