Dansylhydrazid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-(Dimethylamino)naphthalinsulfonsäurehydrazid, 5-(Dimethylamino)naphthalinsulfonohydrazid, 5-(Dimethylamino)naphthalin-1-sulfonohydrazid, Dansylhydrazin

30434-1G 30434-250MG 30434-5G
SIAL30434-1GEA 174 EUR
SIAL30434-1G SIAL30434-250MG SIAL30434-5G
Dansylhydrazid, Sigma-Aldrich®
Dansylhydrazid
Formel: C₁₂H₁₅N₃O₂S
Molecular Weight: 265,34 g/mol
MDL: MFCD00003986
CAS-Nummer: 33008-06-9
EINECS: 251-337-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR