1-Acetylnaphthalin ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1'-Acetonaphthon, Methyl-α-naphthylketon
A12918.0B
A12918.18
A12918.30
A12918.0BEA
286
EUR
A12918.0B
A12918.18
A12918.30
1-Acetylnaphthalin ≥97%
1-Acetylnaphthalin
Purity: 97+%
CAS No.: 941-98-0
Molecular Formula: C12H10O
Formula Weight: 170.21
Flash Point:
Formel:
C₁₂H₁₀O Molecular Weight: 170,21 g/mol Siedepunkt: 302 °C Schmelzpunkt: 9…11 °C Dichte: 1,120 Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004013 CAS-Nummer: 941-98-0 EINECS: 213-384-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...