2-Acetylnaphthalin ≥99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2'-Acetonaphthon, Methyl-β-naphthylketon
A14793.18
A14793.0B
A14793.22
A14793.30
A14793.18EA
21.7
EUR
A14793.18
A14793.0B
A14793.22
A14793.30
2-Acetylnaphthalin ≥99%
2-Acetylnaphthalin
Purity: 99%
CAS No.: 93-08-3
Molecular Formula: C12H10O
Formula Weight: 170.21
Flash Point: 168°(334°F)
Melting Point: 52-56°
Boiling Point: 300-301°
Beilstein Reference: 774965
UN No.: 3077
MDL No.: MFCD00004108
Formel:
C₁₂H₁₀O Molecular Weight: 170,21 g/mol Siedepunkt: 300…301 °C Schmelzpunkt: 52…56 °C Dichte: 1,1171 g/cm³ (20 °C) Flammpunkt: 168 °C (334 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004108 CAS-Nummer: 93-08-3 EINECS: 202-216-2 UN: 3077 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...