Zinkphosphid ≥99,999% (Metall-Basis)
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Trizinkdiphosphid, Zinkphosphid
45459.09
45459.04
45459.02
45459.09EA
1086
EUR
45459.09
45459.04
45459.02
Zinkphosphid ≥99,999% (Metall-Basis)
Zinkphosphid
Purity: 99.999%
CAS No.: 1314-84-7
Molecular Formula: Zn3P2
Formula Weight: 258.10
Physical Form: 6mm & down pieces
Density: 4.55
Melting Point: 420°
Boiling Point: 1100°
Merck Reference: 14,10152
UN No.: 1714
MDL No.: MFCD00049631
Formel:
P₂Zn₃ Molecular Weight: 258,12 g/mol Siedepunkt: 1100 °C Schmelzpunkt: 420 °C Dichte: 4,55 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00049631 CAS-Nummer: 1314-84-7 EINECS: 215-244-5 UN: 1714 ADR: 4.3,I Merck Index: 14,10152 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...