Vanadiumoxytrichlorid (≥28,5% V⁵⁺)

Lieferant: Thermo Scientific
Gefahr

Synonyme: Vanadiumoxidtrichlorid, Vanadyl(V)chlorid, Vanadium(V)oxychlorid, Vanadiumtrichloridoxid, Vanadyltrichlorid, Vanadiumoxychlorid, Trichloroxovanadium, Vanadium(V)trichloridoxid

81118.09 81118.22 81118.36
81118.09EA 20.5 EUR
81118.09 81118.22 81118.36
Vanadiumoxytrichlorid (≥28,5% V⁵⁺)
Vanadiumoxytrichlorid

CAS No.: 7727-18-6 Molecular Formula: VOCl3 Formula Weight: 173.30 Physical Form: Liquid Density: 1.829 Sensitivity: Moisture Sensitive Appearance: Yellow Odour: Chlorine-like Melting Point: -77° Boiling Point: 126-127° Merck Reference: 14,9927 UN No.: 2443 MDL No.: MFCD00011459

Formel: Cl₃OV
Molecular Weight: 173,3 g/mol
Siedepunkt: 126…127 °C
Schmelzpunkt: –77 °C
Dichte: 1,829
Lagertemperatur: Raumtemperatur
MDL: MFCD00011459
CAS-Nummer: 7727-18-6
EINECS: 231-780-2
UN: 2443
ADR: 8,II
Merck Index: 14,9927

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR