Molybdän(VI)oxid ≥99,5% ACS

Lieferant: Thermo Scientific
Gefahr

Synonyme: Molybdenum trioxide

36687.22 36687.36
36687.22EA 56.6 EUR
36687.22 36687.36
Molybdän(VI)oxid ≥99,5% ACS
Molybdenum(VI) oxide

Purity: 99.5% min CAS No.: 1313-27-5 Molecular Formula: MoO3 Formula Weight: 143.94 Physical Form: Powder Density: 4.692 Appearance: White to grey Odour: Odourless Melting Point: 795° Boiling Point: 1155° subl. Solubility: Slightly soluble in water. Soluble in concentrated mineral acids, alkali hydroxide solutions Merck Reference: 14,6239 UN No.: 3288 MDL No.: MFCD00003469


Source of molybdenum compounds, in agriculture, in analytical chemistry, as a corrosion inhibitor, in ceramic glazes, enamels and pigments, as a catalyst

Formel: MoO₃
Molecular Weight: 143,94 g/mol
Siedepunkt: 1155 °C (Subl.)
Schmelzpunkt: 795 °C
Dichte: 4,692
Lagertemperatur: Raumtemperatur
MDL: MFCD00003469
CAS-Nummer: 1313-27-5
EINECS: 215-204-7
UN: 3288
ADR: 6.1,III
Merck Index: 14,6239

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR