Mangan(II)acetylacetonat, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
Mangan(II)-2,4-pentandionat, Bis(2,4-pentandionato-O,O')mangan(II), Mangan(II)-bis(2,4-pentandionato-O,O'), Di(2,4-pentandionato-O,O')mangan(II)
M-Clarity™ Qualitätsstufe = MQ100
245763-100G
245763-5G
SIAL245763-100GEA
140
EUR
SIAL245763-100G
SIAL245763-5G
Mangan(II)acetylacetonat, Sigma-Aldrich®
Mangan(II)acetylacetonat
Formel:
C₁₀H₁₄MnO₄ Molecular Weight: 253,16 g/mol |
MDL:
MFCD00000022 CAS-Nummer: 14024-58-9 EINECS: 237-858-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...