Palladium(II)oxid, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
520748-1G
520748-5G
SIAL520748-1GEA
180
EUR
SIAL520748-1G
SIAL520748-5G
Palladium(II)oxid, Sigma-Aldrich®
Palladium(II) oxide
Formel:
OPd Molecular Weight: 122,42 g/mol Schmelzpunkt: 870 °C Dichte: 8,73 g/cm³ (20 °C) |
MDL:
MFCD00011172 CAS-Nummer: 1314-08-5 EINECS: 215-218-3 Merck Index: 12,07125 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...