Palladium(II)acetylacetonat (34,7% Pd)

Lieferant: Thermo Scientific
Gefahr

Synonyme: Palladium(II)-bis(2,4-pentandionato-O,O'), Bis(2,4-pentandionato-O,O')palladium(II), Di(2,4-pentandionato-O,O')palladium(II), Palladium(II)-2,4-pentandionat

10517.03 10517.06
10517.03EA 112 EUR
10517.03 10517.06
Palladium(II)acetylacetonat (34,7% Pd)
Palladium(II)acetylacetonat

CAS No.: 14024-61-4 Molecular Formula: C10H14O4Pd Formula Weight: 304.92 Physical Form: Powder Appearance: Yellow to orange Odour: Odourless MDL No.: MFCD00000025


Carbonylation, oligomerization

Formel: C₁₀H₁₄O₄Pd
Molecular Weight: 304,64 g/mol
Siedepunkt: ∼190 °C (1013 hPa)
Schmelzpunkt: 210 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00000025
CAS-Nummer: 14024-61-4
EINECS: 237-859-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR