Cobalt(III)acetylacetonat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Tris(2,4-pentandionato-O,O')cobalt(III), Tri(2,4-pentandionato-O,O')cobalt(III), Cobalt(III)-2,4-pentandionat, Cobalt(III)-tris(2,4-pentandionato-O,O')

494534-25G 494534-5G
SIAL494534-25GEA 250 EUR
SIAL494534-25G SIAL494534-5G
Cobalt(III)acetylacetonat, Sigma-Aldrich®
Cobalt(III)acetylacetonat
Formel: C₁₅H₂₁CoO₆
Molecular Weight: 356,26 g/mol
MDL: MFCD00013488
CAS-Nummer: 21679-46-9
EINECS: 244-527-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR