Rhodium(III)acetylacetonat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Rhodium(III)-2,4-pentandionat, Tris(2,4-pentandionato-O,O')rhodium(III), Tri(2,4-pentandionato-O,O')rhodium(III), Rhodium(III)-tris(2,4-pentandionato-O,O')

M-Clarity™ Qualitätsstufe = MQ100

282774-1G 282774-250MG 282774-5G
SIAL282774-1GEA 473 EUR
SIAL282774-1G SIAL282774-250MG SIAL282774-5G
Rhodium(III)acetylacetonat, Sigma-Aldrich®
Rhodium(III)acetylacetonat
Formel: C₁₅H₂₁O₆Rh
Molecular Weight: 400,23 g/mol
Schmelzpunkt: 202…205 °C
MDL: MFCD00083144
CAS-Nummer: 14284-92-5
EINECS: 244-527-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR