Ruthenium(III)acetylacetonat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Tris(2,4-pentandionato-O,O')ruthenium(III), Tri(2,4-pentandionato-O,O')ruthenium(III), Ruthenium(III)-tris(2,4-pentandionato-O,O'), Ruthenium(III)-2,4-pentandionat

M-Clarity™ Qualitätsstufe = MQ100

282766-1G 282766-5G
SIAL282766-1GEA 152 EUR
SIAL282766-1G SIAL282766-5G
Ruthenium(III)acetylacetonat, Sigma-Aldrich®
Ruthenium(III)acetylacetonat
Formel: C₁₅H₂₁O₆Ru
Molecular Weight: 398,4 g/mol
MDL: MFCD00000030
CAS-Nummer: 14284-93-6
EINECS: 238-193-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR