Cobalt(II)sulfid ≥99,5% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Cobaltsulfid
13114.14
13114.22
13114.14EA
89.4
EUR
13114.14
13114.22
Cobalt(II)sulfid ≥99,5% (Metall-Basis)
Cobalt sulphide
Purity: 99.5%
CAS No.: 1317-42-6
Molecular Formula: CoS (generally Co6S5 and/or Co4S3)
Formula Weight: 91.00
Physical Form: -100 Mesh Powder
Odour: Odourless
UN No.: 3077
MDL No.: MFCD00016032
Formel:
CoS Schmelzpunkt: >1116 °C Dichte: 5,45 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016032 CAS-Nummer: 1317-42-6 EINECS: 215-273-3 UN: 3077 ADR: 9,III Merck Index: 12,02511 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...