2-Pyrazincarbonsäure ≥99%

Lieferant: Thermo Scientific

Synonyme: Pyrazincarbonsäure, Pyrazin-2-carbonsäure

A13363.14 A13363.22
A13363.14EA 30 EUR
A13363.14 A13363.22
2-Pyrazincarbonsäure ≥99%
Pyrazine-2-carboxylic acid

Purity: 99% CAS No.: 98-97-5 Molecular Formula: C5H4N2O2 Formula Weight: 124.10 Melting Point: ca 225° dec. Beilstein Reference: 112305 Merck Reference: 14,7959 MDL No.: MFCD00006130

Formel: C₅H₄N₂O₂
Molecular Weight: 124,1 g/mol
Siedepunkt: 313 °C (1013 hPa)
Schmelzpunkt: ∼ 225 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00006130
CAS-Nummer: 98-97-5
EINECS: 202-718-1
Merck Index: 14,7959

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR