2,3-Pyrazindicarbonsäure ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Pyrazin-2,3-dicarbonsäure
B22108.06
B22108.14
B22108.06EA
28.5
EUR
B22108.06
B22108.14
2,3-Pyrazindicarbonsäure ≥98%
Pyrazine-2,3-dicarboxylic acid
Purity: 98%
CAS No.: 89-01-0
Molecular Formula: C6H4N2O4
Formula Weight: 168.11
Melting Point: ca 188° dec.
Beilstein Reference: 147982
Merck Reference: 14,7958
MDL No.: MFCD00006131
Formel:
C₆H₄N₂O₄ Molecular Weight: 168,11 g/mol Schmelzpunkt: ∼ 188 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006131 CAS-Nummer: 89-01-0 EINECS: 201-875-3 Merck Index: 14,7958 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...