2,3-Pyrazindicarbonsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Pyrazin-2,3-dicarbonsäure

M-Clarity™ Qualitätsstufe = MQ100

P56208-100G P56208-25G
SIALP56208-100GEA 109 EUR
SIALP56208-100G SIALP56208-25G
2,3-Pyrazindicarbonsäure, Sigma-Aldrich®
Pyrazine-2,3-dicarboxylic acid
Formel: C₆H₄N₂O₄
Molecular Weight: 168,11 g/mol
MDL: MFCD00006131
CAS-Nummer: 89-01-0
EINECS: 201-875-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR