2-Iodpyrazin 95%
Lieferant: Thermo Scientific
|
Achtung
|
H26244.03
H26244.06
H26244.03EA
51.7
EUR
H26244.03
H26244.06
2-Iodpyrazin 95%
2-Iodopyrazine
Purity: 95%
CAS No.: 32111-21-0
Molecular Formula: C4H3IN2
Formula Weight: 205.98
Flash Point:
Formel:
C₄H₃IN₂ Molecular Weight: 205,99 g/mol Siedepunkt: 109…110 °C (34 mmHg) Dichte: 2,086 Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD01319019 CAS-Nummer: 32111-21-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...