2-Aminopyrazin 99+%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Aminopyrazin, Pyrazin-2-amin
104490100
104490500
ACRO104490100EA
63.5
EUR
ACRO104490100
ACRO104490500
2-Aminopyrazin 99+%
2-Aminopyrazine
Formel:
C₄H₅N₃ Molecular Weight: 95,1 g/mol Siedepunkt: 203 °C (1013 hPa) Schmelzpunkt: 117…120 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006137 CAS-Nummer: 5049-61-6 EINECS: 225-748-7 UN: 1325 ADR: 4.1,III |
Spezifikation Testergebnisse
Appearance | Light yellow to beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 117 to 120 °C |
Titration with HClO4 | ≥99.0 % |
Hydroxyl value | ≥150 mg KOH/g |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...