2-Methoxypyrazin

Lieferant: FLUOROCHEM

002749-25G 002749-5G
FLUO002749-25GEA 45.9 EUR
FLUO002749-25G FLUO002749-5G
2-Methoxypyrazin
2-Methoxypyrazine
Formel: C₅H₆N₂O
Molecular Weight: 110,12 g/mol
Siedepunkt: 147…149 °C (1013 hPa)
Dichte: 1,143 g/cm³ (20 °C)
MDL: MFCD00006126
CAS-Nummer: 3149-28-8
EINECS: 221-579-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR