1-(3-Methyl-2-pyrazinyl)ethanon

Lieferant: FLUOROCHEM

Synonyme: 1-(3-Methylpyrazinyl)ethan-1-on

002742-5G 002742-25G
FLUO002742-5GEA 51.8 EUR
FLUO002742-5G FLUO002742-25G
1-(3-Methyl-2-pyrazinyl)ethanon
2-Acetyl-3-methylpyrazine
Formel: C₇H₈N₂O
Molecular Weight: 136,15 g/mol
Siedepunkt: 90 °C (1013 hPa)
Dichte: 1,11 g/cm³ (20 °C)
MDL: MFCD00014612
CAS-Nummer: 23787-80-6
EINECS: 245-889-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR