4-Nitropyridin-N-oxid ≥97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
4-Nitropyridine N-oxide
B21404.14
B21404.22
B21404.14EA
51.5
EUR
B21404.14
B21404.22
4-Nitropyridin-N-oxid ≥97%
4-Nitropyridin-N-oxid
Purity: 97%
CAS No.: 1124-33-0
Molecular Formula: C5H4N2O3
Formula Weight: 140.10
Melting Point: 158-163°
Beilstein Reference: 124331
UN No.: 2811
MDL No.: MFCD00006206
Formel:
C₅H₄N₂O₃ Molecular Weight: 140,1 g/mol Siedepunkt: 397 °C (1013 hPa) Schmelzpunkt: 158…163 °C Dichte: 1,52 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006206 CAS-Nummer: 1124-33-0 EINECS: 214-395-4 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...