2-Hydroxy-3-nitropyridin

Lieferant: Thermo Scientific

Synonyme: 3-Nitro-2-pyridinol

SPB06066.1 SPB06066.10 SPB06066.50
MAYBSPB06066.1EA 0 EUR
MAYBSPB06066.1 MAYBSPB06066.10 MAYBSPB06066.50
2-Hydroxy-3-nitropyridin
2-Hydroxy-3-nitropyridine
Formel: C₅H₄N₂O₃
Molecular Weight: 140,1 g/mol
Schmelzpunkt: 224 °C
MDL: MFCD00006270
CAS-Nummer: 6332-56-5
EINECS: 228-709-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR