4-Hydroxy-3-nitropyridin 95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3-Nitro-4-pyridinol

B20963.06 B20963.14 B20963.22
B20963.06EA 40.1 EUR
B20963.06 B20963.14 B20963.22
4-Hydroxy-3-nitropyridin 95%
4-Hydroxy-3-nitropyridine

Purity: 95% CAS No.: 5435-54-1 Molecular Formula: C5H4N2O3 Formula Weight: 140.10 Melting Point: ca 280° dec. Beilstein Reference: 1072928

Formel: C₅H₄N₂O₃
Molecular Weight: 140.10 g/mol
Schmelzpunkt: ∼ 280 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00128874
CAS-Nummer: 5435-54-1

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR