3-Amino-2-methylpyridin

Lieferant: FLUOROCHEM

Synonyme: 2-Methylpyridin-3-amin, 3-Amino-2-picolin, 2-Methyl-3-pyridylamin

036315-25G 036315-5G
FLUO036315-25GEA 159 EUR
FLUO036315-25G FLUO036315-5G
3-Amino-2-methylpyridin
3-Amino-2-methylpyridin
Formel: C₆H₈N₂
Molecular Weight: 108.14 g/mol
MDL: MFCD03788195
CAS-Nummer: 3430-10-2
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR