4-Nitro-2-picolin-N-oxid

Lieferant: FLUOROCHEM

Synonyme: 2-Methyl-4-nitropyridin-N-oxid, 2-Methyl-4-nitropyridin-1-oxid, 4-Nitro-2-picolin-N-oxid

003384-1G 003384-5G 003384-25G
FLUO003384-1GEA 32.4 EUR
FLUO003384-1G FLUO003384-5G FLUO003384-25G
4-Nitro-2-picolin-N-oxid
4-Nitro-2-picolin-N-oxid
Formel: C₆H₆N₂O₃
Molecular Weight: 154,13 g/mol
Schmelzpunkt: 156 °C
MDL: MFCD00014625
CAS-Nummer: 5470-66-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR