Cyanursäure
Lieferant: Lovibond
Synonyme:
1,3,5-Triazacyclohexan-2,4,6-trion
511370BT
511371BT
LOVI511370BTEA
25.8
EUR
LOVI511370BT
LOVI511371BT
Cyanursäure
Cyanursäure
Formel:
C₃H₃N₃O₃ Molecular Weight: 129,08 g/mol Dichte: 2,5 g/cm³ (20 °C) |
MDL:
MFCD00082990 CAS-Nummer: 108-80-5 EINECS: 203-618-0 Merck Index: 13,02728 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...