Tris(2-hydroxyethyl)-1,3,5-triazintrion ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1,3,5-Tris(2-hydroxyethyl)isocyanursäure, 1,3,5-Tris(2-hydroxyethyl)isocyanurat, 1,3,5-Tris(2-hydroxyethyl)cyanursäure
B21697.0B
B21697.18
B21697.30
B21697.0BEA
443
EUR
B21697.0B
B21697.18
B21697.30
Tris(2-hydroxyethyl)-1,3,5-triazintrion ≥97%
Tris(2-hydroxyethyl)-1,3,5-triazintrion
Purity: 97%
CAS No.: 839-90-7
Molecular Formula: C9H15N3O6
Formula Weight: 261.24
Melting Point: 136-140°
MDL No.: MFCD00003549
Formel:
C₉H₁₅N₃O₆ Molecular Weight: 261,23 g/mol Siedepunkt: 527 °C (1013 hPa) Schmelzpunkt: 136…140 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003549 CAS-Nummer: 839-90-7 EINECS: 212-660-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...