2,4,6-Trichlor-1,3,5-triazin ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 2,4,6-Trichlor-s-triazin, Cyanurchlorid

L03442.18 L03442.30 L03442.0B
L03442.18EA 24.3 EUR
L03442.18 L03442.30 L03442.0B
2,4,6-Trichlor-1,3,5-triazin ≥98%
2,4,6-Trichloro-1,3,5-triazine

Purity: 98% CAS No.: 108-77-0 Molecular Formula: C3Cl3N3 Formula Weight: 184.41 Storage Temperature: 2°C to 8°C Flash Point: 190°(374°F) Melting Point: 145-149° Boiling Point: 192-194° Beilstein Reference: 124246 UN No.: 2670 MDL No.: MFCD00006046

Formel: C₃Cl₃N₃
Molecular Weight: 184,41 g/mol
Siedepunkt: 192…194 °C
Schmelzpunkt: 145…149 °C
Dichte: 1,920
Flammpunkt: >190 °C (374 °F)
MDL: MFCD00006046
CAS-Nummer: 108-77-0
EINECS: 203-614-9
UN: 2670
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR