1,3,5-Triazin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: s-Triazin

M-Clarity™ Qualitätsstufe = MQ200

T46051-1G T46051-5G
SIALT46051-1GEA 45.6 EUR
SIALT46051-1G SIALT46051-5G
1,3,5-Triazin, Sigma-Aldrich®
1,3,5-Triazine
Formel: C₃H₃N₃
Molecular Weight: 81,08 g/mol
MDL: MFCD00006044
CAS-Nummer: 290-87-9
EINECS: 206-028-1
UN: 2928
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR