1-Adamantancarbonsäure 99%

Lieferant: Thermo Scientific
Achtung

Synonyme: 1-Adamantanecarboxylic acid, Tricyclo[3.3.1.1(3,7)]decane-1-carboxylic acid

147880250 147881000 147880050
ACRO147880250EA 53.2 EUR
ACRO147880250 ACRO147881000 ACRO147880050
1-Adamantancarbonsäure 99%
1-Adamantancarbonsäure
Formel: C₁₁H₁₆O₂
Molecular Weight: 180,25 g/mol
Schmelzpunkt: 172…176 °C
Dichte: 1,221 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00074720
CAS-Nummer: 828-51-3
EINECS: 212-584-6

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow to light beige Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 172 to 176 °C
Titration with NaOH 98.5 to 101.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR