1-Adamantancarbonsäure

Lieferant: Thermo Scientific

Synonyme: 1-Adamantanecarboxylic acid, Tricyclo[3.3.1.1(3,7)]decane-1-carboxylic acid

SB01219.1 SB01219.10 SB01219.50
MAYBSB01219.1EA 0 EUR
MAYBSB01219.1 MAYBSB01219.10 MAYBSB01219.50
1-Adamantancarbonsäure
1-Adamantancarbonsäure
Formel: C₁₁H₁₆O₂
Molecular Weight: 180,25 g/mol
Schmelzpunkt: 172…175 °C
Dichte: 1,221 g/cm³ (20 °C)
MDL: MFCD00074720
CAS-Nummer: 828-51-3
EINECS: 212-584-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR