Eusolex® 2292
Lieferant: Merck
1.05382.0101
1.05382.1001
1.05382.9026
1.05382.0100
1.05382.0101EA
0
EUR
1.05382.0101
1.05382.1001
1.05382.9026
1.05382.0100
Eusolex® 2292
Eusolex®
Organic UV filter
Eusolex® 2292 is a clear, liquid, oil soluble and effective UV-B filter. It's easy to process in an oil phase and shows good solubilizing properties for solid crystalline UV filters. Eusolex® 2292 is worldwide approved.
INCI name: Ethylhexyl Methoxycinnamate, BHT
Formel:
C₁₈H₂₆O₃ Siedepunkt: 160 °C (1 hPa) Dichte: 1,005…1.013 g/cm3 (25 °C) Lagertemperatur: Raumtemperatur |
CAS-Nummer:
5466-77-3 |
Spezifikation Testergebnisse
Color | colourless to light yellow (< 100 Hazen) |
Appearance | clear liquid |
Odour | practical odourless |
Assay Ethylhexyl methoxycinnamate (GC) | 95.0 - 105.0 % |
Chromatography purity | conforms |
Identity (retention time) | conforms |
Identit (IR-spectrum) | conforms |
Refractive index (n 20/D) | 1.542 - 1.548 |
Density (d 20/20) | 1.010 - 1.016 |
Density (d 25/25) | 1.005 - 1.013 |
Spee. absorbance(at 308 nm; 1 %; 1 cm; methanol) | ≥ 830 |
Heavy metals (as Pb) | ≤ 0.002% |
As (Arsenic) | ≤ 0.0002% |
Residual Solvents ICHQ3C | excluded by manufacturing process |
Acidity | conforms |
Sulfated ash | ≤ 0.1 % |
Water | ≤ 0.5 % |
Microbial Purity (Total Viable Aerobic Count) | ≤ 100 CFU/g |
E.coli | absent in 1 g |
Pseudomonas aeruginosa | absent in 1 g |
Staphylococcus aureus | absent in 1 g |
Candida albicans | absent in 1 g |
Salmonella species | absent in 10 g |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...