2-tert-Butyldimethylsiloxybut-3-yne 97%, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: tert-Butyl-dimethyl-(methyl-prop-2-ynloxy)silane

M-Clarity™ Qualitätsstufe = MQ600

667579-10G 667579-1G
SIAL667579-10GEA 248 EUR
SIAL667579-10G SIAL667579-1G
2-tert-Butyldimethylsiloxybut-3-yne 97%, Sigma-Aldrich®
2-tert-Butyldimethylsiloxybut-3-yne
Formel: C₁₀H₂₀OSi
Molecular Weight: 184,35 g/mol
Flammpunkt: 39 °C
MDL: MFCD08276883
CAS-Nummer: 125494-93-1

Bestellen

Spezifikation Testergebnisse

Appearance (Colour) Colourless to Yellow
Appearance (Form) Liquid
Infrared spectrum Conforms to Structure
Purity (GC) ≥ 96.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR